missing translation for 'onlineSavingsMsg'
Learn More

3-Maleimidopropionic acid N-hydroxysuccinimide ester, 99%

Product Code. 15497675
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15497675 250 mg 250mg
11382419 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497675 Supplier Thermo Scientific Alfa Aesar Supplier No. H27329.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 55750-62-4 | C11H10N2O6 | 266.209 g/mol

3-Maleimidopropionic acid N-hydroxysuccinimide ester is a reagent that is widely used for generating stable maleimide-activated carrier proteins that can spontaneously react with sulfhydryls. Alternatively these relatively stable maleimide-activated intermediates may be lyophilized and stored for later conjugation to a hapten.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Maleimidopropionic acid N-hydroxysuccinimide ester is a reagent that is widely used for generating stable maleimide-activated carrier proteins that can spontaneously react with sulfhydryls. Alternatively these relatively stable maleimide-activated intermediates may be lyophilized and stored for later conjugation to a hapten.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55750-62-4
Molecular Formula C11H10N2O6
Molecular Weight (g/mol) 266.209
MDL Number MFCD00043141
InChI Key JKHVDAUOODACDU-UHFFFAOYSA-N
Synonym n-succinimidyl 3-maleimidopropionate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoate, 3-maleimido propionic acid n-hydroxysuccinimide ester, bmps, 3-maleimidopropionic acid n-succinimidyl ester, 3-maleimidopropionic acid nhs, succinimido 3-maleimidopropanoate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl propanoate, 3-maleimidopropionic acid n-hydroxysuccinimide ester
PubChem CID 4620597
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 3-(2,5-dioxopyrrol-1-yl)propanoate
SMILES C1CC(=O)N(C1=O)OC(=O)CCN2C(=O)C=CC2=O

Specifications

Melting Point 168°C to 170°C
Quantity 250 mg
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 266.21
Percent Purity 99%
Chemical Name or Material 3-Maleimidopropionic acid N-hydroxysuccinimide ester
compliance-icons
Product Identifier
  • 3-Maleimidopropionic acid N-hydroxysuccinimide ester
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 3-Maleimidopropionicacid N-hydroxysuccinimide ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.