missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-1,2-propanediol, 90 wt% aqueous solution

Product Code. 13408338 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
This item is not returnable. View return policy

Product Code. 13408338

Brand: Thermo Scientific Acros 125525000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-27-5 | C3H8O2S | 108.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-27-5, 7732-18-5
Molecular Formula C3H8O2S
Molecular Weight (g/mol) 108.16
MDL Number MFCD00004879
InChI Key PJUIMOJAAPLTRJ-UHFFFAOYNA-N
Synonym 3-mercapto-1,2-propanediol, monothioglycerol, 1-thioglycerol, thioglycerol, thioglycerine, thioglycerin, thiovanol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiol
PubChem CID 7291
ChEBI CHEBI:74537
IUPAC Name 3-sulfanylpropane-1,2-diol
SMILES OCC(O)CS

Specifications

CAS Min % 88.0, 8.0
CAS Max % 92.0, 12.0
Color Colorless to Yellow
Density 1.2300g/mL
Boiling Point 118.0°C (5.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula HSCH2CH(OH)CH2OH
Refractive Index 1.5073
Quantity 500 mL
Beilstein 01,519
Merck Index 15,9488
Specific Gravity 1.23
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol,insoluble in ether
Formula Weight 108.16
Percent Purity ≥88%
Physical Form Viscous Solution
Chemical Name or Material 3-Mercapto-1, 2-propanediol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.