missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-2-butanone, 98%, stab. with 0.1% Calcium carbonate

Product Code. 11420687
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420687 25 g 25g
11410687 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420687 Supplier Thermo Scientific Alfa Aesar Supplier No. B22494.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40789-98-8 | C4H8OS | 104.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 40789-98-8
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.17
MDL Number MFCD00085206
InChI Key XLMPYCGSRHSSSX-UHFFFAOYNA-N
Synonym 3-mercapto-2-butanone, 2-butanone, 3-mercapto, 2-mercapto-3-butanone, 3-mercaptobutan-2-one, fema no. 3298, mercapto-3 butanone-2 french, mercapto-3 butanone-2, acmc-209jgq, 4-01-00-03993 beilstein handbook reference
PubChem CID 38679
IUPAC Name 3-sulfanylbutan-2-one
SMILES CC(S)C(C)=O

Specifica

Density 1.015
Boiling Point 50°C (18 mmHg)
Flash Point 47°C (117°F)
Refractive Index 1.475
Quantity 25 g
UN Number UN1224
Sensitivity Light sensitive
Formula Weight 104.17
Percent Purity 98%
Chemical Name or Material 3-Mercapto-2-butanone, stabilized with 0.1% Calcium carbonate

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.