missing translation for 'onlineSavingsMsg'
Learn More

3-Mercaptopropionic acid, 99%

Product Code. 11450803
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450803 500 g 500g
11440803 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450803 Supplier Thermo Scientific Alfa Aesar Supplier No. A13261.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 107-96-0 | C3H6O2S | 106.139 g/mol

3-Mercaptopropionic acid is widely used in food and beverage industries as a flavoring agent. It is used in the production of PVC stabilizers, which are used as chain transfer agents in polymerizations. It can be used as primary or secondary, color stabilizer in combination with phenolic antioxidant for polymers. It acts as a sulfide ion equivalent and is utilized in the preparation of diaryl sulfide from aryl iodide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Mercaptopropionic acid is widely used in food and beverage industries as a flavoring agent. It is used in the production of PVC stabilizers, which are used as chain transfer agents in polymerizations. It can be used as primary or secondary, color stabilizer in combination with phenolic antioxidant for polymers. It acts as a sulfide ion equivalent and is utilized in the preparation of diaryl sulfide from aryl iodide.

Solubility
Miscible with water, alcohol, benzene and ether.

Notes
Air Sensitive. Hygroscopic. Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-96-0
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.139
MDL Number MFCD00004897
InChI Key DKIDEFUBRARXTE-UHFFFAOYSA-N
Synonym 3-mercaptopropionic acid, 3-mercaptopropanoic acid, 3-thiopropionic acid, 3-thiopropanoic acid, propanoic acid, 3-mercapto, beta-mercaptopropionic acid, 3mpa, mercaptopropionic acid, beta-thiopropionic acid, 2-mercaptoethanecarboxylic acid
PubChem CID 6514
ChEBI CHEBI:44111
IUPAC Name 3-sulfanylpropanoic acid
SMILES C(CS)C(=O)O

Especificaciones

Melting Point 17°C to 19°C
Density 1.22
Boiling Point 110°C to 111°C (15 mmHg)
Flash Point 93°C (199°F)
Linear Formula HSCH2CH2CO2H
Refractive Index 1.492
Quantity 500 g
UN Number UN2922
Beilstein 773807
Solubility Information Miscible with water,alcohol,benzene and ether.
Formula Weight 106.14
Percent Purity 99%
Chemical Name or Material 3-Mercaptopropionic acid
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • 3-Mercaptopropionic acid
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Indicaciones de peligro
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Otros peligros
  • MIXTURE LIST-Contain: 3-Mercaptopropionicacid

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.