Learn More
(3-Mercaptopropyl)trimethoxysilane, 95%
Description
(3-Mercaptopropyl)trimethoxysilane is used as a capping molecule for cadmium sulphide nanocrystals. It is used in the preparation of multi-walled carbon nanotubes and bilayer modified gold electrode. It acts as a linker molecule used in the preparation of gold multilayers and as a molecular adhesive in the fabrication of vapor-deposited gold electrodes on glass substrates. It is also used as a precursor for silica.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(3-Mercaptopropyl)trimethoxysilane is used as a capping molecule for cadmium sulphide nanocrystals. It is used in the preparation of multi-walled carbon nanotubes and bilayer modified gold electrode. It acts as a linker molecule used in the preparation of gold multilayers and as a molecular adhesive in the fabrication of vapor-deposited gold electrodes on glass substrates. It is also used as a precursor for silica.
Solubility
Miscible with methanol, ethanol, isopropanol, acetone, benzene, toluene and xylene.
Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, strong acids, alcohols and water. Decomposes in water.
Specifications
Specifications
| Boiling Point | 213°C to 215°C |
| Melting Point | <-50°C |
| Quantity | 25 g |
| Linear Formula | (CH3O)3Si(CH2)3SH |
| UN Number | UN3082 |
| Beilstein | 2038119 |
| Flash Point | 96°C (204°F) |
| Solubility Information | Miscible with methanol,ethanol,isopropanol,acetone,benzene,toluene and xylene. |
| Formula Weight | 196.34 |
| Percent Purity | 95% |
| Show More |
Safety and Handling
- (3-Mercaptopropyl)trimethoxysilane
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- MIXTURE LIST-Contain: 3-Mercaptopropyltrimethoxysilane
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.