missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-1,2-propanediol, 98%

Product Code. 10214851 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10214851 1 g Glass bottle 1g
10557431 5 g Glass bottle 5g
10791145 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10214851 Supplier Thermo Scientific Acros Supplier No. 338390010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 623-39-2 | C4H10O3 | 106.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-39-2
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.12
MDL Number MFCD00004719
InChI Key PSJBSUHYCGQTHZ-UHFFFAOYNA-N
Synonym 3-methoxy-1,2-propanediol, 1,2-propanediol, 3-methoxy, 1-o-methylglycerol, glycerol 1-monomethyl ether, glycerin-alpha-monomethyl ether, glycerin-.alpha.-monomethyl ether, glycerin methyl ether, 3-methoxypropan-1,2-diol, 1-o-methyl-rac-glycerol, acmc-1bhm7
PubChem CID 92822
IUPAC Name 3-methoxypropane-1,2-diol
SMILES COCC(O)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.1200g/mL
Boiling Point 133°C (35.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H3COCH2CH(OH)CH2OH
Refractive Index 1.4430 to 1.445
Quantity 1 g
Specific Gravity 1.12
Solubility Information Solubility in water: soluble in water
Formula Weight 106.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methoxy-1, 2-propanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.