missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-1-butanol, 99%

Product Code. 10295170 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10295170 1 L Glass bottle 1 L
10255220 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10295170 Supplier Thermo Scientific Acros Supplier No. 125790010
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2517-43-3 | C5H12O2 | 104.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2517-43-3
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00002931
InChI Key JSGVZVOGOQILFM-UHFFFAOYNA-N
Synonym 3-methoxy-1-butanol, 3-methoxybutanol, 1-butanol, 3-methoxy, methoxybutanol, ccris 8976, 1-butanol, 3-methoxy-,-, 1-butanol, 3-methoxy-, r, 3-methoxy-butanol, 3-methoxybutane-1-ol, 3-methoxybutyl alcohol
PubChem CID 17291
IUPAC Name 3-methoxybutan-1-ol
SMILES COC(C)CCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -85.0°C
Color Colorless
Density 0.9200g/mL
Boiling Point 161.0°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OCH3)CH2CH2OH
Refractive Index 1.4150 to 1.4165
Quantity 1 L
UN Number 1987
Specific Gravity 0.92
Solubility Information Solubility in water: soluble. Other solubilities: miscible with most common organic solvents
Formula Weight 104.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Methoxy-1-butanol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methoxy-1-butanol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.