missing translation for 'onlineSavingsMsg'
Learn More

3-[Methoxy(methyl)carbamoyl]benzeneboronic acid, 97%, Alfa Aesar™

Product Code. 15450616
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450616 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15450616 Supplier Thermo Scientific Alfa Aesar Supplier No. H53354.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 723281-57-0
Molecular Formula C9H12BNO4
Molecular Weight (g/mol) 209.01
MDL Number MFCD04115699
InChI Key IYZAWDLXZOCCSB-UHFFFAOYSA-N
Synonym 3-n,o-dimethylhydroxylaminocarbonyl phenylboronic acid, 3-methoxy methyl carbamoyl phenyl boronic acid, 3-methoxy methyl carbamoyl phenylboronic acid, 3-borono-n-methoxy-n-methylbenzamide, n-methoxy-n-methyl 3-boronobenzamide, 3-n,o-dimethylhydroxylaminocarbonyl benzeneboronic acid, 3-n,o-dimethyl hydroxylaminocarbonyl phenylboronic acid, 3-n ,o-dimethylhydroxylamino-carbonyl phenylboronic acid
PubChem CID 3856172
IUPAC Name [3-[methoxy(methyl)carbamoyl]phenyl]boronic acid
SMILES CON(C)C(=O)C1=CC=CC(=C1)B(O)O

Specifications

Melting Point 66°C to 70°C
Quantity 1 g
Formula Weight 209.01
Percent Purity 97%
Chemical Name or Material 3-[Methoxy(methyl)carbamoyl]benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.