missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-p-tyramine Hydrochloride, TRC

Product Code. 17982675
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17982675 100 mg 100mg
17914905 500 mg 500mg
17903785 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17982675 Supplier Toronto Research Chemicals Supplier No. TRCM332320100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1477-68-5
Molecular Formula C9 H13 N O2 . Cl H
Molecular Weight (g/mol) 203.67
Synonym Phenol, 4-(2-aminoethyl)-2-methoxy-, hydrochloride (1:1), Guaiacol, 4-(2-aminoethyl)-, hydrochloride (6CI), Phenol, 4-(2-aminoethyl)-2-methoxy-, hydrochloride (8CI,9CI), 2-Methoxytyramine hydrochloride, 3-Methoxy-4-hydroxyphenethylamine hydrochloride, 3-O-Methyldopamine hydrochloride, 4-(2-Aminoethyl)-2-methoxyphenol hydrochloride, 4-Hydroxy-3-methoxy-β-phenethylamine hydrochloride, 4-Hydroxy-3-methoxyphenethylamine hydrochloride, 3-Methoxy-p-tyramine hydrochloride
IUPAC Name 4-(2-aminoethyl)-2-methoxyphenol;hydrochloride
SMILES Cl.COc1cc(CCN)ccc1O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H13NO2.ClH/c1-12-9-6-7(4-5-10)2-3-8(9)11;/h2-3,6,11H,4-5,10H2,1H3;1H
Formula Weight 203.0713
Percent Purity >95%
Chemical Name or Material 3-Methoxy-p-tyramine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.