missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyazetidine hydrochloride, 95%

Product Code. 15424198
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424198 500 mg 500mg
15414198 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15424198 Supplier Thermo Scientific Alfa Aesar Supplier No. H57769.ME
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 148644-09-1 | C4H10ClNO | 123.58 g/mol

It is used in the preparation of isoxazole-thiazole derivatives as GABA. It is a receptor inverse agonists for use in the treatment of cognitive disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of isoxazole-thiazole derivatives as GABA. It is a receptor inverse agonists for use in the treatment of cognitive disorders.

Notes
Air sensitive. Store in a cool ventilated place. Avoid strong oxidizing materials and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148644-09-1
Molecular Formula C4H10ClNO
Molecular Weight (g/mol) 123.58
MDL Number MFCD06804514
InChI Key KSXGQRBTBLQJEF-UHFFFAOYSA-N
Synonym 3-methoxyazetidine hydrochloride, 3-methoxy-azetidine hydrochloride, 3-azetidinyl methyl ether hydrochloride, 3-methoxyazetidine hcl, azetidine, 3-methoxy-, hydrochloride, azetidine, 3-methoxy-,hydrochloride 1:1, acmc-20a0d9, ksc495e1t, 3-methoxyazetidinehydrochloride, 3-methoxy-azetidine-hydrochloride
PubChem CID 22242858
IUPAC Name 3-methoxyazetidine;hydrochloride
SMILES COC1CNC1.Cl

Specifications

Boiling Point 143°C to 149°C
Quantity 500 mg
Formula Weight 123.58
Percent Purity 95%
Chemical Name or Material 3-Methoxyazetidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.