missing translation for 'onlineSavingsMsg'
Learn More

3-(Methoxycarbonyl)benzeneboronic acid, 97%

Product Code. 11387289
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387289 1 g 1g
11397289 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387289 Supplier Thermo Scientific Alfa Aesar Supplier No. H27444.03
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 99769-19-4 | C8H9BO4 | 179.97 g/mol

Reagent used for tandem-type Pd(II)-catalyzed oxidative Heck reaction and intramolecular C-H amidation sequence, copper-mediated ligandless aerobic fluoroalkylation of arylboronic acids with fluoroalkyl iodides, one-pot ipso-nitration of arylboronic acids, copper-catalyzed nitration, cyclocondensation followed by palladium-phosphine-catalyzed Suzuki-Miyaura coupling. Reagent used in Preparation of biaryls via nickel-catalyzed Suzuki-Miyaura cross-coupling reaction of aryl halides with arylboronic acid6, chromenones and their bradykinin B1 antagonistic active, Pt nanoparticles at Photoactive metal-organic frameworks resulting in efficient hydrogen evolution via synergistic photoexcitation and electron injecti, salicylate-based thienylbenzoic acids as E. coli methionine aminopeptidase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent used for tandem-type Pd(II)-catalyzed oxidative Heck reaction and intramolecular C-H amidation sequence, copper-mediated ligandless aerobic fluoroalkylation of arylboronic acids with fluoroalkyl iodides, one-pot ipso-nitration of arylboronic acids, copper-catalyzed nitration, cyclocondensation followed by palladium-phosphine-catalyzed Suzuki-Miyaura coupling. Reagent used in Preparation of biaryls via nickel-catalyzed Suzuki-Miyaura cross-coupling reaction of aryl halides with arylboronic acid6, chromenones and their bradykinin B1 antagonistic active, Pt nanoparticles at Photoactive metal-organic frameworks resulting in efficient hydrogen evolution via synergistic photoexcitation and electron injecti, salicylate-based thienylbenzoic acids as E. coli methionine aminopeptidase inhibitor.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99769-19-4
Molecular Formula C8H9BO4
Molecular Weight (g/mol) 179.97
MDL Number MFCD02093046
InChI Key ALTLCJHSJMGSLT-UHFFFAOYSA-N
Synonym 3-methoxycarbonyl phenylboronic acid, 3-methoxycarbonylphenyl boronic acid, 3-methoxycarbonyl phenyl boronic acid, 3-methoxycarbonyl benzeneboronic acid, 3-methoxycarbonylphenylbaronic acid, methyl 3-dihydroxyboranyl benzoate, m-methoxycarbonyl phenylboronic acid, 3-carbomethoxy-phenylboronic acid
PubChem CID 2734714
SMILES COC(=O)C1=CC=CC(=C1)B(O)O

Specifications

Melting Point 136°C
Quantity 1 g
Solubility Information Reacts with water.
IUPAC Name [3-(methoxycarbonyl)phenyl]boronic acid
Formula Weight 179.97
Percent Purity 97%
Chemical Name or Material 3-(Methoxycarbonyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.