missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylacetic Acid, TRC

Product Code. 30467241
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30467241 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30467241 Supplier Toronto Research Chemicals Supplier No. TRCM2657705G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Methoxyphenylacetic Acid
CAS 1798-09-0
Molecular Formula C9 H10 O3
Quantity 5 g
Synonym Benzeneacetic acid','3-methoxy-, Acetic acid','(m-methoxyphenyl)- (6CI,7CI,8CI), 3-Methoxybenzeneacetic acid, (3-Methoxyphenyl)acetic acid, (m-Methoxyphenyl)acetic acid, 2-(3-Methoxyphenyl)acetic acid, [3-(Methyloxy)phenyl]acetic acid, m-Anisylacetic acid, 3-Methoxyphenylacetic acid
InChI Formula InChI=1S/C9H10O3/c1-12-8-4-2-3-7(5-8)6-9(10)11/h2-5H,6H2,1H3,(H,10,11)
SMILES COc1cccc(CC(=O)O)c1
IUPAC Name 2-(3-methoxyphenyl)acetic acid
Molecular Weight (g/mol) 166.17
Formula Weight 166.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.