missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylmagnesium bromide, 1.0 M in 2-MeTHF

Product Code. 15431586
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15431586 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15431586 Supplier Thermo Scientific Alfa Aesar Supplier No. H54780.AE
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 36282-40-3 | C7H7BrMgO | 211.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 36282-40-3
Molecular Formula C7H7BrMgO
MDL Number MFCD00672002
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Synonym 3-methoxyphenylmagnesium bromide, 3-methoxyphenyl magnesium bromide, 3-methoxyphenylmagnesium bromide solution, 3-methoxyphenylmagnesiumbromide, 3-methoxyphenylmagnesiumbromid, bromo 3-methoxyphenyl magnesium, 3-methoxy phenylmagnesium bromide, 3-methoxyphenyl-magnesium bromide, m-methoxyphenyl magnesium bromide, 3-methoxylphenyl magnesium bromide, grignard reagent
InChI Key AJKNYEHIJAMGHX-UHFFFAOYSA-M
SMILES COC1=CC=CC([Mg]Br)=C1
IUPAC Name magnesium(2+) anisole bromide
Molecular Weight (g/mol) 211.34
PubChem CID 4605969
Formula Weight 211.34
Concentration or Composition (by Analyte or Components) 1.0M in 2-MeTHF
Chemical Name or Material 3-Methoxyphenylmagnesium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.