missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypropyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 10327504
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10327504 10 g 10g
10001894 50 g 50g
10729425 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10327504 Supplier Thermo Scientific Maybridge Supplier No. TL04220EA
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 17702-11-3
Molecular Formula C5H9NOS
Molecular Weight (g/mol) 131.193
MDL Number MFCD00022063
InChI Key UKVFWFDXYBLOLU-UHFFFAOYSA-N
Synonym 3-methoxypropyl isothiocyanate, 3-methoxypropylisothiocyanate, 1-isothiocyanato-3-methoxy-propane, propane,1-isothiocyanato-3-methoxy, propane, 1-isothiocyanato-3-methoxy, 3-methoxypropanisothiocyanate, acmc-1bu13, 3-methoxy-n-propyl isothiocyanate, 3-methoxpropyl isothiocyanate, #
PubChem CID 140274
IUPAC Name 1-isothiocyanato-3-methoxypropane
SMILES COCCCN=C=S

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 131.2
Percent Purity 97%
Chemical Name or Material 3-Methoxypropyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.