missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypropyl isothiocyanate, 98%, Thermo Scientific™

Product Code. 11472238
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472238 5 g 5g
11482238 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11472238 Supplier Thermo Scientific Alfa Aesar Supplier No. A15216.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17702-11-3
Molecular Formula C5H9NOS
Molecular Weight (g/mol) 131.193
MDL Number MFCD00022063
InChI Key UKVFWFDXYBLOLU-UHFFFAOYSA-N
Synonym 3-methoxypropyl isothiocyanate, 3-methoxypropylisothiocyanate, 1-isothiocyanato-3-methoxy-propane, propane,1-isothiocyanato-3-methoxy, propane, 1-isothiocyanato-3-methoxy, 3-methoxypropanisothiocyanate, acmc-1bu13, 3-methoxy-n-propyl isothiocyanate, 3-methoxpropyl isothiocyanate, #
PubChem CID 140274
IUPAC Name 1-isothiocyanato-3-methoxypropane
SMILES COCCCN=C=S

Specifications

Density 1.03
Boiling Point 119°C to 120°C (35 mmHg)
Flash Point 77°C (170°F)
Odor Pungent
Refractive Index 1.503
Quantity 5 g
UN Number UN2922
Beilstein 1751281
Sensitivity Moisture sensitive
Formula Weight 131.2
Percent Purity 98%
Chemical Name or Material 3-Methoxypropyl isothiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.