missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypropylamine, 99%

Product Code. 11401107
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11401107 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11401107 Supplier Thermo Scientific Alfa Aesar Supplier No. B24095.AK
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 5332-73-0 | C4H11NO | 89.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5332-73-0
Melting Point <-70°C
Density 0.872
Boiling Point 117°C to 119°C
Flash Point 27°C (81°F)
Odor Amine-like
Molecular Formula C4H11NO
Refractive Index 1.4175
MDL Number MFCD00014831
Quantity 250 mL
UN Number UN2734
Beilstein 878144
Sensitivity Air sensitive; Hygroscopic
Synonym 3-methoxypropylamine, 1-propanamine, 3-methoxy, 1-amino-3-methoxypropane, 3-aminopropyl methyl ether, 3-methyoxypropylamine, 3-methoxy-n-propylamine, 3-methoxy-1-propanamine, propylamine, 3-methoxy, 3-mpa, propanolamine methyl ether
InChI Key FAXDZWQIWUSWJH-UHFFFAOYSA-N
SMILES COCCCN
IUPAC Name 3-methoxypropan-1-amine
Molecular Weight (g/mol) 89.14
PubChem CID 1672
Formula Weight 89.14
Percent Purity 99%
Chemical Name or Material 3-Methoxypropylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Methoxypropylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 3-Methoxypropylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.