missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypropylamine, 99+%, Thermo Scientific Chemicals

Product Code. 10533411 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10533411 1 L Glass bottle 1 L
10714353 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10533411 Supplier Thermo Scientific Acros Supplier No. 257520010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5332-73-0
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00014831
InChI Key FAXDZWQIWUSWJH-UHFFFAOYSA-N
Synonym 3-methoxypropylamine, 1-propanamine, 3-methoxy, 1-amino-3-methoxypropane, 3-aminopropyl methyl ether, 3-methyoxypropylamine, 3-methoxy-n-propylamine, 3-methoxy-1-propanamine, propylamine, 3-methoxy, 3-mpa, propanolamine methyl ether
PubChem CID 1672
IUPAC Name 3-methoxypropan-1-amine
SMILES COCCCN

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -65°C
Color Colorless
Density 0.8700g/mL
Boiling Point 118°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3OCH2CH2CH2NH2
Refractive Index 1.4165 to 1.4185
Quantity 1 L
Specific Gravity 0.87
Solubility Information Solubility in water: miscible. Other solubilities: miscible with ethanol,toluene,acetone,ether,,carbon tetrachloride and hexane
Viscosity 0.91 mPa.s (20°C)
Formula Weight 89.12
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 3-Methoxypropylamine, 99+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Methoxypropylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.