missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxythiophene-2-carboxaldehyde, 97%

Product Code. 15476827
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476827 250 mg 250mg
15466827 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476827 Supplier Thermo Scientific Alfa Aesar Supplier No. H33940.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35134-07-7 | C6H6O2S | 142.17 g/mol

3-Methoxythiophene-2-carboxaldehyde is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxythiophene-2-carboxaldehyde is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from air, oxidizing agents. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35134-07-7
Molecular Formula C6H6O2S
Molecular Weight (g/mol) 142.17
MDL Number MFCD01571098
InChI Key KGJDTMQUUPIAEF-UHFFFAOYSA-N
PubChem CID 588315
IUPAC Name 3-methoxythiophene-2-carbaldehyde
SMILES COC1=C(SC=C1)C=O

Specifications

Melting Point 80°C to 82°C
Quantity 250 mg
Sensitivity Air Sensitive
Synonym 3-methoxy-2-thiophenecarboxaldehyde, 3-methoxythiophene-2-carboxaldehyde, 2-thiophenecarboxaldehyde, 3-methoxy, 3-methoxy-2-thenaldehyde, 2-formyl-3-methoxythiophene, 3-methoxy-2-thiophenealdehyde, 3-methoxy-2-thiophenecarbaldehyde, 3-methoxy-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,3-methoxy, 3-methoxy-2-thiophenecarbaldehyde #
Solubility Information Slightly soluble in water.
Formula Weight 142.18
Percent Purity 97%
Chemical Name or Material 3-Methoxythiophene-2-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.