missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-butanethiol, TRC

Product Code. 30481530
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481530 10 g 10g
30472923 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30481530 Supplier Toronto Research Chemicals Supplier No. TRCM28889510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-31-1
Molecular Formula C5H12S
Molecular Weight (g/mol) 104.21
Synonym 2-Methyl-4-butanethiol, 3-Methylbutanethiol, 3-Methylbutyl Mercaptan, Isoamyl Mercaptan, Isoamyl Sulfhydrate, Isopentanethiol, Isopentyl Mercaptan, Isopentylthiol, iso-Amyl Mercaptan
IUPAC Name 3-methylbutane-1-thiol
SMILES CC(C)CCS

Specifications

Quantity 10 g
InChI Formula InChI=1S/C5H12S/c1-5(2)3-4-6/h5-6H,3-4H2,1-2H3
Formula Weight 104.066
Chemical Name or Material 3-Methyl-1-butanethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.