missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-butanol, ACS reagent

Product Code. 10228623 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10228623 2.5 L Glass bottle 2.5 L
10517752 500 mL Glass bottle 500mL
10330244 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10228623 Supplier Thermo Scientific Acros Supplier No. 412730025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-51-3 | C5H12O | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-51-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002934
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Synonym 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
PubChem CID 31260
ChEBI CHEBI:15837
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO

Specifications

Melting Point -70.0°C
Density 0.8000g/mL
Boiling Point 130.0°C
Flash Point 45°C
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CH2OH
Refractive Index 1.4050 to 1.4070
Quantity 2.5 L
Beilstein 01, 392
Residue after Evaporation 0.003% max.
Merck Index 15, 5241
Specific Gravity 0.8
Solubility Information Solubility in water: 25g/L (20°C). Other solubilities: miscible with alcohol,ether,benzene,oils,,chloroform
Viscosity 4.3 mPa.s (20°C)
Formula Weight 88.15
Percent Purity ≥98.5%
Grade ACS Reagent
Titratable Acid 0.002 meq/g max
Physical Form Liquid
Chemical Name or Material 3-Methyl-1-butanol, 98.5%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Methyl-1-butanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.