missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-butyne, 96%

Product Code. 15470248
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470248 25 g 25g
15460248 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15470248 Supplier Thermo Scientific Alfa Aesar Supplier No. H53439.14
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 598-23-2 | C5H8 | 68.119 g/mol

3-Methyl-1-butyne is a terminal alkyne that under goes hydrosilylation reaction using phenylsilane. 3-Methyl-1-butyne is used as an intermediate for pharmaceuticals, in cosmetics and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-1-butyne is a terminal alkyne that under goes hydrosilylation reaction using phenylsilane. 3-Methyl-1-butyne is used as an intermediate for pharmaceuticals, in cosmetics and in chemical research.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Store in cool place. Light and heat sensitive. Incompatible with strong oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-23-2
Molecular Formula C5H8
Molecular Weight (g/mol) 68.119
MDL Number MFCD00039853
InChI Key USCSRAJGJYMJFZ-UHFFFAOYSA-N
Synonym 3-methyl-1-butyne, 1-butyne, 3-methyl, isopentyne, isopropylacetylene, isopropyl acetylene, 3-methylbutyne, 2-methyl-3-butyne, 3-methyl-but-1-yne, 3-methyl,1-butyne
PubChem CID 69019
ChEBI CHEBI:87379
IUPAC Name 3-methylbut-1-yne
SMILES CC(C)C#C

Specifications

Density 0.666
Boiling Point 30°C
Flash Point −30°C (−22°F)
Linear Formula (CH3)2CHC≡CH
Refractive Index 1.374
Quantity 25 g
UN Number UN3295
Solubility Information Miscible with alcohol. Immiscible with water.
Formula Weight 68.12
Percent Purity 97%
Chemical Name or Material 3-Methyl-1-butyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.