missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-butyne, 96%

Product Code. 10698214 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
Packaging:
Glass bottle
Unit Size:
10mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10698214 10 mL Glass bottle 10mL
10457493 1 mL Glass bottle 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10698214 Supplier Thermo Scientific Acros Supplier No. 392740100
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 598-23-2 | C5H8 | 68.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-23-2
Molecular Formula C5H8
Molecular Weight (g/mol) 68.12
MDL Number MFCD00039853
InChI Key USCSRAJGJYMJFZ-UHFFFAOYSA-N
Synonym 3-methyl-1-butyne, 1-butyne, 3-methyl, isopentyne, isopropylacetylene, isopropyl acetylene, 3-methylbutyne, 2-methyl-3-butyne, 3-methyl-but-1-yne, 3-methyl,1-butyne
PubChem CID 69019
ChEBI CHEBI:87379
IUPAC Name 3-methylbut-1-yne
SMILES CC(C)C#C

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.6660g/mL
Boiling Point 29.5°C
Flash Point -30°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHC∽CH
Refractive Index 1.3730 to 1.3750 (20°C, 589nm)
Quantity 10 mL
Specific Gravity 0.666
Formula Weight 68.12
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Methyl-1-butyne
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Methyl-1-butyne
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H224-Extremely flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.