missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-phenyl-1H-pyrazole, 98%

Product Code. 11409130
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409130 1 g 1g
11429130 10 g 10g
11419130 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409130 Supplier Thermo Scientific Alfa Aesar Supplier No. A14841.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1128-54-7 | C10H10N2 | 158.20 g/mol

3-Methyl-1-phenyl-1H-pyrazole is used to produce 3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde at the temperature of 90-100°C with reagent phosphorus oxychloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-1-phenyl-1H-pyrazole is used to produce 3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde at the temperature of 90-100°C with reagent phosphorus oxychloride.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1128-54-7
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.20
MDL Number MFCD00003123
InChI Key RJXLUGSJEMSDPK-UHFFFAOYSA-N
PubChem CID 70783
IUPAC Name 3-methyl-1-phenylpyrazole
SMILES CC1=NN(C=C1)C1=CC=CC=C1

Specifications

Melting Point 35°C to 38°C
Flash Point >110°C (230°F)
Odor Odorless
Quantity 1 g
Beilstein 117870
Synonym 3-methyl-1-phenyl-1h-pyrazole, 1-phenyl-3-methylpyrazole, pyrazole, 3-methyl-1-phenyl, 1h-pyrazole, 3-methyl-1-phenyl, 1h-pyrazole, methylphenyl, 1h-pyrazole,3-methyl-1-phenyl, acmc-20an11, ksc494g4n, rjxlugsjemsdpk-uhfffaoysa, 3-methyl-1-phenyl-1h-pyrazole #
Solubility Information Insoluble in water.
Formula Weight 158.2
Percent Purity 98%
Chemical Name or Material 3-Methyl-1-phenyl-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.