missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-phenyl-1H-pyrazole, 98%, Thermo Scientific™

Product Code. 11355306 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11355306 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11355306 Supplier Thermo Scientific Acros Supplier No. 295390025
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1128-54-7
CAS Min % 97.5
CAS Max % 100.0
Melting Point 33°C to 37°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C10H10N2
MDL Number MFCD00003123
Quantity 2.5 g
Synonym 3-methyl-1-phenyl-1h-pyrazole, 1-phenyl-3-methylpyrazole, pyrazole, 3-methyl-1-phenyl, 1h-pyrazole, 3-methyl-1-phenyl, 1h-pyrazole, methylphenyl, 1h-pyrazole,3-methyl-1-phenyl, acmc-20an11, ksc494g4n, rjxlugsjemsdpk-uhfffaoysa, 3-methyl-1-phenyl-1h-pyrazole #
InChI Key RJXLUGSJEMSDPK-UHFFFAOYSA-N
SMILES CC1=NN(C=C1)C1=CC=CC=C1
IUPAC Name 3-methyl-1-phenylpyrazole
Molecular Weight (g/mol) 158.20
PubChem CID 70783
Formula Weight 158.2
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material 3-Methyl-1-phenyl-1H-pyrazole, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-1-phenyl-1H-pyrazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.