missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-phenyl-2-butanone, 96%

Product Code. 15448869
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448869 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15448869 Supplier Thermo Scientific Alfa Aesar Supplier No. L07176.06
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2893-05-2 | C6H15NO | 117.19 g/mol

3-Methyl-1-phenyl-2-butanone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-1-phenyl-2-butanone is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 2893-05-2
Density 0.968
Boiling Point 237°C to 240°C
Molecular Formula C6H15NO
Refractive Index 1.504
MDL Number MFCD00040487
Quantity 5 g
Beilstein 2042880
Synonym 3-isopropoxypropylamine, 3-isopropoxy-1-propylamine, isopropoxypropylamine, 1-propanamine, 3-1-methylethoxy, 3-aminopropyl isopropyl ether, 3-1-methylethoxy-1-propanamine, 3-isopropoxy-n-propylamine, 3-isopropoxypropan-1-amine, propylamine, 3-isopropoxy, 3-propan-2-yloxy propan-1-amine
Solubility Information Not miscible in water.
InChI Key VHYUNSUGCNKWSO-UHFFFAOYSA-N
SMILES CC(C)OCCCN
IUPAC Name 3-propan-2-yloxypropan-1-amine
Molecular Weight (g/mol) 117.19
PubChem CID 76194
Formula Weight 162.23
Percent Purity 96%
Chemical Name or Material 3-Methyl-1-phenyl-2-butanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.