missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1H-pyrazole-5-carboxylic acid, 97%

Product Code. 15496338
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15496338 1 g 1g
15406348 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496338 Supplier Thermo Scientific Alfa Aesar Supplier No. H59899.03
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 402-61-9 | C5H6N2O2 | 126.115 g/mol

3-Methylpyrazole-5-carboxylic acid is a potent and selective D-amino acid oxidase (DAO) (also known as DAAO, DAMOX and OXDA) inhibitor that protects DAO cells from oxidative stress induced by D-Serine. 3-Methylpyrazole-5-carboxylic acid specifically prevents formalin-induced tonic pain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylpyrazole-5-carboxylic acid is a potent and selective D-amino acid oxidase (DAO) (also known as DAAO, DAMOX and OXDA) inhibitor that protects DAO cells from oxidative stress induced by D-Serine. 3-Methylpyrazole-5-carboxylic acid specifically prevents formalin-induced tonic pain.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-61-9
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
MDL Number MFCD00462235
InChI Key WSMQKESQZFQMFW-UHFFFAOYSA-N
Synonym 3-methyl-1h-pyrazole-5-carboxylic acid, 5-methylpyrazole-3-carboxylic acid, 5-methyl-2h-pyrazole-3-carboxylic acid, 3-methylpyrazole-5-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-methyl, 5-methylpyrazole-3-carboxylicacid, 3-methyl-1h-pyrazole-5-carboxylicacid, pyrazole-3-carboxylic acid, 5-methyl, pubchem7842, 5-methyl-2h-pyrazole-3-carboxylicacid
PubChem CID 9822
ChEBI CHEBI:74739
IUPAC Name 5-methyl-1H-pyrazole-3-carboxylic acid
SMILES CC1=CC(=NN1)C(=O)O

Specifications

Melting Point 236°C to 240°C
Quantity 1 g
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material 3-Methyl-1H-pyrazole-5-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.