missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1H-pyrazole, 97%

Product Code. 11351347
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11351347 5 g 5g
11361347 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11351347 Supplier Thermo Scientific Alfa Aesar Supplier No. L01618.06
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1453-58-3 | C4H6N2 | 82.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1453-58-3
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005240,MFCD08685900
InChI Key XKVUYEYANWFIJX-UHFFFAOYSA-N
PubChem CID 15073
IUPAC Name 5-methyl-1H-pyrazole
SMILES CC1=CC=NN1

Specifications

Density 1.02
Boiling Point 203°C to 205°C
Flash Point 93°C (199°F)
Refractive Index 1.496
Quantity 5 g
Beilstein 1454
Synonym 3-methylpyrazole, 3-methyl-1h-pyrazole, 1h-pyrazole, 3-methyl, 5-methylpyrazole, 3 5-methylpyrazole, pyrazole, 5-methyl, 1h-pyrazole,5-methyl, 1h-pyrazole, 5-methyl, pyrazole, 3-methyl, unii-w7ku1rro6u
Formula Weight 82.11
Percent Purity 97%
Chemical Name or Material 3-Methyl-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.