missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate, 98+%

Product Code. 15482436
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15482436 50 g 50g
15472436 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15482436 Supplier Thermo Scientific Alfa Aesar Supplier No. H56061.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 38894-11-0 | C8H12ClN3OS | 233.714 g/mol

It is used as a reagent for the spectrophotometric determination of aliphatic aldehydes and for the determination of hexosamines in glycosaminoglycans. It is Incorporated into a new peroxidase color reaction. It is also a reagent for the spectrophotometric determination of traces of selenium (IV) in environmental samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent for the spectrophotometric determination of aliphatic aldehydes and for the determination of hexosamines in glycosaminoglycans. It is Incorporated into a new peroxidase color reaction. It is also a reagent for the spectrophotometric determination of traces of selenium (IV) in environmental samples.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38894-11-0
Molecular Formula C8H12ClN3OS
Molecular Weight (g/mol) 233.714
MDL Number MFCD00149370
InChI Key IYXXQOGEFHAQGU-PIHABLKOSA-N
Synonym 2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride hydrate, 3-methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate, 3-methyl-2-benzothiazolinone hydrazone hcl monohydrate, 3-methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, e-3-methyl-1,3-benzothiazol-2-ylidene hydrazine hydrate hydrochloride, e-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride hydrate, 2z-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrate hydrochloride
PubChem CID 6508671
IUPAC Name (E)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrate;hydrochloride
SMILES CN1C2=CC=CC=C2SC1=NN.O.Cl

Specifications

Melting Point 270°C to 274°C
Quantity 50 g
UN Number UN2811
Beilstein 4722243
Solubility Information Soluble in water.
Formula Weight 233.72 (215.70 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material 3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate
compliance-icons
Product Identifier
  • 3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.