missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-butanone, 98%

Product Code. 10338450 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
250mL
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Packaging unitSize
10338450 250 mL Glass bottle 250mL
10042990 1 L Glass bottle 1 L
10475755 5 L Plastic drum 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10338450 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 218772500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 563-80-4 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 563-80-4
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00008919
InChI Key SYBYTAAJFKOIEJ-UHFFFAOYSA-N
Synonym 3-methyl-2-butanone, methyl isopropyl ketone, isopropyl methyl ketone, 2-acetylpropane, 2-butanone, 3-methyl, methyl butanone-2, mipk, 3-methylbutanone, ketone, isopropyl methyl, 2-methyl-3-butanone
PubChem CID 11251
IUPAC Name 3-methylbutan-2-one
SMILES CC(C)C(C)=O

Especificaciones

CAS Min % 97.5
CAS Max % 100.0
Melting Point -92.0°C
Color Colorless
Density 0.8000g/mL
Boiling Point 94.0°C to 95.0°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCOCH3
Refractive Index 1.3870 to 1.3890
Quantity 250 mL
Beilstein 01, 682
Merck Index 15, 6162
Specific Gravity 0.8
Solubility Information Solubility in water: 6g/L (20 c). Other solubilities: miscible with most common organic solvents
Formula Weight 86.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-butanone
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 3-Methyl-2-butanone
Palabra de advertencia
  • Danger
Clasificación
  • Flammable liquid Category 2
Indicaciones de peligro
  • H225-Highly flammable liquid and vapour.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.