missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-butanone, puriss, ≥98.5% (GC), Honeywell™

Product Code. 15695890 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15695890 100 mL Glass bottle 100mL
15605900 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15695890 Supplier Honeywell Research Chemicals Supplier No. 59600100ML
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 563-80-4
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00008919
InChI Key SYBYTAAJFKOIEJ-UHFFFAOYSA-N
Synonym 3-methyl-2-butanone, methyl isopropyl ketone, isopropyl methyl ketone, 2-acetylpropane, 2-butanone, 3-methyl, methyl butanone-2, mipk, 3-methylbutanone, ketone, isopropyl methyl, 2-methyl-3-butanone
PubChem CID 11251
IUPAC Name 3-methylbutan-2-one
SMILES CC(C)C(C)=O

Specifications

Melting Point -92°C
Color Colorless
Density 0.805g/mL at 25°C
Boiling Point 94°C to 95°C
Packaging Glass bottle
Linear Formula (CH3)2CHCOCH3
Refractive Index 1.388
Quantity 100 mL
UN Number UN2397
Beilstein 1071238
Formula Weight 86.13g/mol
Percent Purity ≥98.5% (GC)
Chemical Name or Material 3-Methyl-2-butanone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.