missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-butenal, 97%

Product Code. 11426410
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426410 25 g 25g
11416410 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11426410 Supplier Thermo Scientific Alfa Aesar Supplier No. B22226.14
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 107-86-8 | C5H8O | 84.118 g/mol

3-Methyl-2-butenal is used in small quantities as food flavor in alcoholic beverages, chewing gums, confection frosting, frozen dairy, fruit ice, gelatin pudding, hard and soft candy and jam jelly. Used in daily flavor, and cosmetic flavor in perfumes. A starting material for the synthesis of vitamin A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-butenal is used in small quantities as food flavor in alcoholic beverages, chewing gums, confection frosting, frozen dairy, fruit ice, gelatin pudding, hard and soft candy and jam jelly. Used in daily flavor, and cosmetic flavor in perfumes. A starting material for the synthesis of vitamin A.

Solubility
Soluble in water (110g/L).

Notes
Air sensitive. Store away from strong oxidizing agents, air, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-86-8
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00010291
InChI Key SEPQTYODOKLVSB-UHFFFAOYSA-N
Synonym 3-methyl-2-butenal, prenal, 3-methylcrotonaldehyde, senecialdehyde, 3,3-dimethylacrolein, senecioaldehyde, 2-butenal, 3-methyl, beta,beta-dimethylacrolein, crotonaldehyde, 3-methyl, beta-methylcrotonaldehyde
PubChem CID 61020
ChEBI CHEBI:15825
IUPAC Name 3-methylbut-2-enal
SMILES CC(=CC=O)C

Specifications

Melting Point -20°C
Density 0.88
Boiling Point 136°C
Flash Point 37°C (99°F)
Odor Characteristic
Linear Formula (CH3)2C=CHCHO
Refractive Index 1.4613
Quantity 25 g
UN Number UN2929
Beilstein 1734740
Sensitivity Air sensitive
Merck Index 14,8448
Solubility Information Soluble in water (110g/L).
Formula Weight 84.12
Percent Purity 97%
Chemical Name or Material 3-Methyl-2-butenal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.