missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-butenal, 97%

Product Code. 10660893 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packaging unitSize
10660893 5 mL Glass bottle 5mL
10455872 25 mL Glass bottle 25mL
11904781 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 10660893 Lieferant Thermo Scientific Acros Lieferanten-Nr. 370180050
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 107-86-8 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 107-86-8
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
InChI Key SEPQTYODOKLVSB-UHFFFAOYSA-N
Synonym 3-methyl-2-butenal, prenal, 3-methylcrotonaldehyde, senecialdehyde, 3,3-dimethylacrolein, senecioaldehyde, 2-butenal, 3-methyl, beta,beta-dimethylacrolein, crotonaldehyde, 3-methyl, beta-methylcrotonaldehyde
PubChem CID 61020
ChEBI CHEBI:15825
IUPAC Name 3-methylbut-2-enal
SMILES CC(=CC=O)C

Spezifikation

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8800g/mL
Boiling Point 132.0°C to 133.0°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C=CHCHO
Refractive Index 1.4610 to 1.4630
Quantity 5 mL
Beilstein 01,IV,3464
Merck Index 15,8587
Specific Gravity 0.88
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol and ether
Formula Weight 84.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-butenal
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • 3-Methyl-2-butenal
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Gefahrenhinweis
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Ergänzende Informationen
  • MIXTURE LIST-Contain: 3-Methyl-2-butenal

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.