missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-butenal, 97%

Product Code. 11904781 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass Bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11904781 100 mL Glass Bottle 100mL
10660893 5 mL Glass Bottle 5mL
10455872 25 mL Glass Bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11904781 Supplier Thermo Scientific Acros Supplier No. 370181000
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 107-86-8 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-86-8
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
InChI Key SEPQTYODOKLVSB-UHFFFAOYSA-N
Synonym 3-methyl-2-butenal, prenal, 3-methylcrotonaldehyde, senecialdehyde, 3,3-dimethylacrolein, senecioaldehyde, 2-butenal, 3-methyl, beta,beta-dimethylacrolein, crotonaldehyde, 3-methyl, beta-methylcrotonaldehyde
PubChem CID 61020
ChEBI CHEBI:15825
IUPAC Name 3-methylbut-2-enal
SMILES CC(=CC=O)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8800g/mL
Boiling Point 132.0°C to 133.0°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3)2C=CHCHO
Refractive Index 1.4610 to 1.4630
Quantity 100 mL
Beilstein 01,IV,3464
Merck Index 15,8587
Specific Gravity 0.88
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol and ether
Formula Weight 84.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-butenal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.