missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-methyloxy-4-benzyloxy-butanoic Acid Methyl Ester, TRC

Product Code. 30477714
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477714 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477714 Supplier Toronto Research Chemicals Supplier No. TRCM26359025MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Methyl-3-methyloxy-4-benzyloxy-butanoic Acid Methyl Ester
CAS 1797024-70-4
Molecular Formula C14H20O4
Quantity 25 mg
Synonym 3-Methyl-3-methyloxy-4-(phenylmethoxy)-butanoic Acid Methyl Ester
InChI Formula InChI=1S/C14H20O4/c1-14(17-3,9-13(15)16-2)11-18-10-12-7-5-4-6-8-12/h4-8H,9-11H2,1-3H3
SMILES O=C(OC)CC(OC)(C)COCC1=CC=CC=C1
IUPAC Name methyl 4-(benzyloxy)-3-methoxy-3-methylbutanoate
Molecular Weight (g/mol) 252.31
Formula Weight 252.14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.