missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitro-1H-pyrazole, 97%

Product Code. 15413997
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15413997 25 g 25g
15403997 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15413997 Supplier Thermo Scientific Alfa Aesar Supplier No. H30860.14
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5334-39-4 | C4H5N3O2 | 127.10 g/mol

3-Methyl-4-nitropyrazole is a reactant used in the preparation of aminopyrazole derivatives as potent, selective and brain penetrant Leucine-rich repeat kinase 2(LRRK2) small molecule inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-4-nitropyrazole is a reactant used in the preparation of aminopyrazole derivatives as potent, selective and brain penetrant Leucine-rich repeat kinase 2(LRRK2) small molecule inhibitors.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5334-39-4
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00037864
InChI Key WTZYTQJELOHMMJ-UHFFFAOYSA-N
Synonym 3-methyl-4-nitro-1h-pyrazole, 3-methyl-4-nitropyrazole, 1h-pyrazole, 3-methyl-4-nitro, 3-methyl-4-nitro pyrazole, 3-me-4-no2 pyrazole, 5-methyl-4-nitropyrazole, acmc-209l6a, ksc497o4j, 4-nitro-5-methyl-1h-pyrazole, 3-methyl-4-nitro-1h-pyrazole #
PubChem CID 79255
IUPAC Name 5-methyl-4-nitro-1H-pyrazole
SMILES CC1=C(C=NN1)[N+]([O-])=O

Specifications

Melting Point 131°C to 134°C
Boiling Point 325°C
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 127.1
Percent Purity 97%
Chemical Name or Material 3-Methyl-4-nitro-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.