missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitropyridine N-oxide, 98%, Thermo Scientific™

Product Code. 10700771 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10700771 1 g Glass bottle 1g
10197753 5 g Glass bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10700771

Brand: Thermo Scientific Acros 308990010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1074-98-2
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00014626
InChI Key SSOURMYKACOBIV-UHFFFAOYSA-N
Synonym 3-methyl-4-nitropyridine n-oxide, 3-methyl-4-nitropyridine 1-oxide, 4-nitro-3-picoline n-oxide, 3-methyl-4-nitropyridine-n-oxide, 3-methyl-4-nitropyridin-1-ium-1-olate, 3-methyl-4-nitropyridine-1-oxide, 3-methyl-4-nitro-pyridine 1-oxide, 4-nitro-3-picoline 1-oxide, 4-nitro-3-picoline-n-oxide, pyridine, 3-methyl-4-nitro-, 1-oxide
PubChem CID 70633
IUPAC Name 3-methyl-4-nitro-1-oxidopyridin-1-ium
SMILES CC1=C(C=C[N+](=C1)[O-])[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 135°C to 139°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: slightly soluble
Formula Weight 154.13
Percent Purity 98%
Physical Form Crystalline Needles or Powder
Chemical Name or Material 3-Methyl-4-nitropyridine N-oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.