missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-phenyl-1H-pyrazole, 97%

Product Code. 11357196
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357196 5 g 5g
11347196 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357196 Supplier Thermo Scientific Alfa Aesar Supplier No. L07250.06
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13788-84-6 | C10H10N2 | 158.20 g/mol

3-Methyl-4-phenyl-1H-pyrazole is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-4-phenyl-1H-pyrazole is used as intermediates.

Solubility
Soluble in DMSO, Methanol. Insoluble in water.

Notes
Protect from heat. Store away from air and oxidizing agents. Incompatible with heat and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13788-84-6
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.20
MDL Number MFCD00051752
InChI Key XTXZCNATVCIKTR-UHFFFAOYSA-N
Synonym 3-methyl-4-phenyl-1h-pyrazole, 3-methyl-4-phenylpyrazole, 3-methyl-4-phenylpyrazol, 3-methyl-4-phenyl-2h-pyrazole, acmc-1brbb, maybridge1_003313, 5-methyl-4-phenylpyrazole, pyrazole-based inhibitor 1, 3-methyl-4-phenyl-1h-pyrazol, 4-phenyl-5-methyl-1h-pyrazole
PubChem CID 139601
IUPAC Name 5-methyl-4-phenyl-1H-pyrazole
SMILES CC1=C(C=NN1)C1=CC=CC=C1

Specifications

Melting Point 140°C to 144°C
Quantity 5 g
Beilstein 3330
Solubility Information Soluble in DMSO,Methanol. Insoluble in water.
Formula Weight 158.2
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Methyl-4-phenyl-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.