missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-methoxycarbonyl-1-benzyl-4-piperidone hydrochloride, 99%, Thermo Scientific™

Product Code. 10378910 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10378910 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10378910 Supplier Thermo Scientific Acros Supplier No. 268090010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 205692-65-5
CAS Min % 98.5
CAS Max % 100.0
Melting Point 190°C to 192°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C15H19NO3·HCl
MDL Number MFCD00143175
Quantity 1 g
Synonym 3-methyl-5-methoxycarbonyl-1-benzyl-4-piperidone hydrochloride, methyl 1-benzyl-5-methyl-4-oxopiperidine-3-carboxylate hydrochloride
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
InChI Key UQGAOMLRURDKSU-UHFFFAOYSA-N
SMILES CC1CN(CC(C1=O)C(=O)OC)CC2=CC=CC=C2.Cl
IUPAC Name methyl 1-benzyl-5-methyl-4-oxopiperidine-3-carboxylate;hydrochloride
Molecular Weight (g/mol) 297.77
PubChem CID 71650946
Formula Weight 297.77
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3-Methyl-5-methoxycarbonyl-1-benzyl-4-piperidone hydrochloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.