missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-phenyl-4-isoxazolecarbaldehyde, 95%, Thermo Scientific™

Product Code. 10203072
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10203072 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10203072 Supplier Thermo Scientific Maybridge Supplier No. CC20304DA
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 89479-66-3
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C11H9NO2
MDL Number MFCD03086122
Quantity 1 g
Synonym 3-methyl-5-phenyl-4-isoxazolecarbaldehyde, 3-methyl-5-phenylisoxazole-4-carbaldehyde, 3-methyl-5-phenyl-isoxazole-4-carbaldehyde, 4-isoxazolecarboxaldehyde,3-methyl-5-phenyl, acmc-20e4b4, 4-formyl-3-methyl-5-phenylisoxazole, 3-methyl-5-phenyl-4-isoxazolecarboxaldehyde
InChI Key IZULDLZFUWWFHF-UHFFFAOYSA-N
SMILES CC1=NOC(=C1C=O)C2=CC=CC=C2
IUPAC Name 3-methyl-5-phenyl-1,2-oxazole-4-carbaldehyde
Molecular Weight (g/mol) 187.198
PubChem CID 2776516
Formula Weight 187.2
Percent Purity 95%
Chemical Name or Material 3-Methyl-5-phenyl-4-isoxazolecarbaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.