missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-d3-4-nitrophenol-2,5,6-d3, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 3-Methyl-4-nitrophenol methyl,ring-d6 |
| CAS | 1219803-89-0 |
| Molecular Formula | C7 2H6 H N O3 |
| Quantity | 0.1 g |
| Synonym | 3-Methyl-4-nitrophenol-D6, 3-(Methyl-d3)-4-nitrophenol-2,5,6-d3, 2-Nitro-5-hydroxytoluene-d6, 4-Nitro-m-cresol-d6, 5-Hydroxy-2-nitrotoluene-α,α,α,3,4,6-d6, 5-Methyl-4-nitrophenol-d6 |
| InChI Formula | InChI=1 S/C7H7NO3/c1-5-4-6(9)2-3-7(5)8(10)11/h2-4,9 H,1H3/i1D3,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(c(c([2 H])c1O)C([2 H])([2 H])[2 H])[N+](=O)[O-] |
| IUPAC Name | 2,3,6-trideuterio-4-nitro-5-(trideuteriomethyl)phenol |
| Molecular Weight (g/mol) | 159.17 |
| Formula Weight | 159.0803 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction