missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-diazirine-3-propanoic Acid, TRC

Product Code. 30482825
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482825 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482825 Supplier Toronto Research Chemicals Supplier No. TRCM299550250MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Methyl-diazirine-3-propanoic Acid
CAS 25055-86-1
Molecular Formula C5 H8 N2 O2
Quantity 250 mg
Synonym 3H-Diazirine-3-propanoic acid','3-methyl-, 3H-Diazirine-3-propionic acid','3-methyl- (8CI), 3-Methyl-3H-diazirine-3-propanoic acid, 3-(3-Methyl-3H-diaziren-3-yl)propanoic acid, 3-(3-Methyldiazirin-3-yl)propanoic acid, 3-Methyl-diazirine-3-propanoic acid
InChI Formula InChI=1S/C5H8N2O2/c1-5(6-7-5)3-2-4(8)9/h2-3H2,1H3,(H,8,9)
SMILES CC1(CCC(=O)O)N=N1
IUPAC Name 3-(3-methyldiazirin-3-yl)propanoic acid
Molecular Weight (g/mol) 128.13
Formula Weight 128.0586
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.