missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-L-phenylalanine, 95%

Product Code. 15448537
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448537 1 g 1g
15468537 250 mg 250mg
15458537 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15448537 Supplier Thermo Scientific Alfa Aesar Supplier No. H52047.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 114926-37-3 | C10H13NO2 | 179.219 g/mol

It finds its application in the identification of N-acyl 4-(5-pyrimidine-2,4-dionyl)phenylalanine derivatives and their orally active prodrug esters as dual-acting alpha4-beta1 and alpha4-beta7 receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the identification of N-acyl 4-(5-pyrimidine-2,4-dionyl)phenylalanine derivatives and their orally active prodrug esters as dual-acting alpha4-beta1 and alpha4-beta7 receptor antagonists.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114926-37-3
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00044972
InChI Key JZRBSTONIYRNRI-VIFPVBQESA-N
Synonym 3-methyl-l-phenylalanine, l-3-methylphenylalanine, 3-methylphenyl-l-alanine, 3-methy-l-phenylalanine, s-2-amino-3-m-tolyl propanoic acid, 3-methylphenylalanine, l-3-methylphe, phenylalanine, 3-methyl, 2s-2-amino-3-3-methylphenyl propanoic acid, l-3-me-phe-oh
PubChem CID 2761494
IUPAC Name (2S)-2-amino-3-(3-methylphenyl)propanoic acid
SMILES CC1=CC(=CC=C1)CC(C(=O)O)N

Specifications

Solubility Information Slightly soluble in water.
Quantity 1 g
Formula Weight 179.22
Percent Purity 95%
Chemical Name or Material 3-Methyl-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.