missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ 3-Methyladenine

Product Code. 18094562
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18094562 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18094562 Supplier R&D Systems™ Supplier No. 3977/50
  • ONLINE EXCLUSIVE PRICE
    189.27 €
    169.05 € / 50mg
    Save 20.22 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Class III PI 3-kinase inhibitor; also inhibits autophagy

Inhibitor of class III phosphatidylinositol 3-kinase (PI 3-kinase); also inhibits the autophagic sequestration of cell proteins in rat hepatocytes. Blocks apoptosis in cerebellar granule cells (CGCs) following serum and potassium deprivation.
TRUSTED_SUSTAINABILITY

Specifications

CAS 5142-23-4
Quantity 50 mg
Product Line Tocris
Molecular Formula C6H7N5
Formula Weight 149.15
Purity >99%
Molecular Weight (g/mol) 149.16
Chemical Name or Material 3-Methyladenine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.