missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies 3-Methyladenine

Product Code. 17100352
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17100352 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17100352 Supplier STEMCELL Technologies Supplier No. 74222
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PI3K/AKT pathway inhibitor

  • 3-Methyladenine has been reported to inhibit autophagy (Seglen & Gordon) by blocking autophagosome formation via the persistent inhibition of class I and transient inhibition of class III phosphoinositide 3-kinase (PI3K; Wu et al.) METABOLISM
  • Inhibits autophagy in isolated rat hepatocytes (Wu et al.)

Specifications

CAS 5142-23-4
Synonym 3-MA; NSC 66389
Quantity 50 mg
Molecular Formula C6H7N5
Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.