missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylamino)oxetane, 95%

Product Code. 15433898
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15433898 250 mg 250mg
15443898 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15433898 Supplier Thermo Scientific Alfa Aesar Supplier No. H57425.MD
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 952182-03-5 | C4H9NO | 87.122 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Very Soluble (307 g/L) (25°C)

Notes
Store at 2-8°C. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 952182-03-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.122
MDL Number MFCD08544349
InChI Key JUIXJPRSYHSLHK-UHFFFAOYSA-N
Synonym n-methyl-3-oxetanamine, 1-methyl-3-oxetanamine, 3-methylamino oxetane, n-methyl-3-oxetaneamine, 3-methylaminooxetane, n-methyloxetane-3-amine, 3-oxetanamine,n-methyl, n-methyl-3-aminooxetane, 3-oxetanamine, n-methyl
PubChem CID 46839993
IUPAC Name N-methyloxetan-3-amine
SMILES CNC1COC1

Specifications

Flash Point 40°C (104°F)
Quantity 250 mg
UN Number UN1993
Solubility Information Very Soluble (307g/L) (25°C)
Formula Weight 87.12
Percent Purity 95%
Chemical Name or Material 3-(Methylamino)oxetane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.