missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylamino)oxetane, 95%

Produktkod. 15443898
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
500 mg
Förpackningsstorlek:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Förpackningsstorlek
15443898 500 mg 500mg
15433898 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15443898 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' H57425.ME
 Restricted Item
Estimated Shipment: 30-10-2026
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 952182-03-5 | C4H9NO | 87.122 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Very Soluble (307 g/L) (25°C)

Notes
Store at 2-8°C. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 952182-03-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.122
MDL Number MFCD08544349
InChI Key JUIXJPRSYHSLHK-UHFFFAOYSA-N
Synonym n-methyl-3-oxetanamine, 1-methyl-3-oxetanamine, 3-methylamino oxetane, n-methyl-3-oxetaneamine, 3-methylaminooxetane, n-methyloxetane-3-amine, 3-oxetanamine,n-methyl, n-methyl-3-aminooxetane, 3-oxetanamine, n-methyl
PubChem CID 46839993
IUPAC Name N-methyloxetan-3-amine
SMILES CNC1COC1

Specifikationer

Flash Point 40°C (104°F)
Quantity 500 mg
UN Number UN1993
Solubility Information Very Soluble (307g/L) (25°C)
Formula Weight 87.12
Percent Purity 95%
Chemical Name or Material 3-(Methylamino)oxetane

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.