missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzo[b]furan-2-carboxylic acid, 97%

Product Code. 11460137
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460137 1 g 1g
11470137 5 g 5g
11480137 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460137 Supplier Thermo Scientific Alfa Aesar Supplier No. B21044.03
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 24673-56-1 | C10H8O3 | 176.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24673-56-1
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
MDL Number MFCD00060511
InChI Key YMZTUCZCQMQFMK-UHFFFAOYSA-N
Synonym 3-methylbenzofuran-2-carboxylic acid, 2-benzofurancarboxylic acid, 3-methyl, 3-methyl-2-benzofurancarboxylic acid, 3-methyl-benzofuran-2-carboxylic acid, 3-methylbenzo b furan-2-carboxylic acid, cambridge id 5100668, acmc-1cp97, 3-methylbenzofuran-2-carboxylicacid, 3-methylbenzofuran 2 carboxylic acid, 2-benzofurancarboxylicacid, 3-methyl
PubChem CID 600591
IUPAC Name 3-methyl-1-benzofuran-2-carboxylic acid
SMILES CC1=C(OC2=CC=CC=C12)C(=O)O

Specifications

Melting Point 192°C to 197°C
Quantity 1 g
Beilstein 132104
Formula Weight 176.17
Percent Purity 97%
Chemical Name or Material 3-Methylbenzo[b]furan-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.