missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Methylbenzyl mercaptan, 95%

Product Code. 11328585
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328585 5 g 5g
11318585 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328585 Supplier Alfa Aesar™ Supplier No. L12043.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 25697-56-7
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00022075
InChI Key PNAZUQUHTIOEHF-UHFFFAOYSA-N
Synonym 3-methylphenyl methanethiol, 3-methylbenzyl mercaptan, m-methylbenzyl mercaptan, 3-methylbenzenemethanethiol, m-methylbenzylmercaptan, acmc-1cq7k, 3-methyl-benzenemethanethiol, benzenemethanethiol,3-methyl, benzenemethanethiol, 3-methyl, 3-methylphenyl methane-1-thiol
PubChem CID 2801207
IUPAC Name (3-methylphenyl)methanethiol
SMILES CC1=CC(=CC=C1)CS

Specifications

Boiling Point 102°C to 104°C (15 mmHg)
Quantity 5 g
UN Number UN2810
Beilstein 2239962
Formula Weight 138.23
Percent Purity 95%
Chemical Name or Material 3-Methylbenzyl mercaptan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.