missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbut-2-enyl formate, Thermo Scientific™

Product Code. 10533222
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10533222 1 g Amber Glass Bottle 1g
10743063 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10533222 Supplier Thermo Scientific Maybridge Supplier No. RDP00092DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 68480-28-4
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00036614
InChI Key ZCROFVOAWLRGFY-UHFFFAOYSA-N
Synonym prenyl formate, 2-buten-1-ol, 3-methyl-, formate, unii-77xfk1mol6, 2-buten-1-ol, 3-methyl-, 1-formate, 3-methylbut-2-en-1-yl formate, 77xfk1mol6, formic acid, 3-methyl-2-butenyl ester, prenylformiat, 3-methyl but-2-enyl formate, 3-methyl-2-butenyl formate #
PubChem CID 110373
SMILES CC(C)=CCOC=O

Specifications

Packaging Amber Glass Bottle
Linear Formula (CH3)2C=CHCH2OCHO
Quantity 1 g
IUPAC Name 3-methylbut-2-en-1-yl formate
Formula Weight 114.14
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-Methylbut-2-enyl formate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.