missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbutanol, Solstice™

Product Code. 16625873
Change view
Click to view available options
Quantity:
200 L
Packaging:
Returnable Stainless Steel Drum
Unit Size:
200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
16625873 200 L Returnable Stainless Steel Drum 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16625873 Supplier Fluka Supplier No. 59090200L
  • ONLINE EXCLUSIVE PRICE
    14520.00 €
    10738.10 € / 200 L
    Save 3781.90 €
    26% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, ≥98.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Specifications

CAS 123-51-3
Boiling Point 131°C to 132°C
Flash Point 43 °C
Packaging Returnable Stainless Steel Drum
Quantity 200 L
PubChem CID 31260
ChEBI CHEBI:15837
Percent Purity min. 98.5 %
Grade ACS General Use Solvents
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.